Learn More
Enzo Life Sciences PKC epsilon (human), (recombinant) (GST-tag) (5 µg)
SDP
Supplier:  Enzo Life Sciences BMLSE4430005
Encompass_Preferred
Alternative name: Protein kinase C epsilon. Formulation: Liquid. In 50mM TRIS-HCl, pH 7.5, containing 150mM NaCl, 0.25mM DTT, 0.1mM EGTA, 0.1mM EDTA, 0.1mM PMSF and 25% glycerol. . Source: Produced in insect cells. Full length active PKCε (protein kinase C ε). Produced in a baculovirus expression system. Long Term Storage: -80°C.
Catalog No. 50-201-2030