Learn More
Cayman Chemical PKCε Inhibitor Scramble Peptide, Purity: ≥95%, CAS: 813416-34-1, 1 mg
SDP
Catalog No. 502978873
Encompass_Preferred
A negative control for PKCε inhibitor peptide; a scrambled peptide with identical amino acid composition to PKCε inhibitor peptide; intended for use as a negative control for PKCε inhibitor peptide activity
Catalog No. 50-297-8873 Supplier Cayman Chemical Supplier No. 317611 MG
A negative control for PKCε inhibitor peptide; a scrambled peptide with identical amino acid composition to PKCε inhibitor peptide; intended for use as a negative control for PKCε inhibitor peptide activity