Learn More
Cayman Chemical PKCβ Inhibitor, Purity: ≥98%, CAS: 257879-35-9, 5 mg
SDP
Catalog No. 502913622
Encompass_Preferred
A PKCβ Inhibitor (IC50s = 21 and 5 nM, respectively); selective for PKCβ1 and PKCβ2 over PKCα, PKCε, and PKCγ (IC50s = 331, 2,807, and >1,000 nM, respectively), as well as JAK1, JAK2, and TYK2 (IC50s = 770, 3,850, and 2,310 nM, respectively); inhibits JAK3 (IC50 = 17 nM); induces apoptosis and inhibits cell cycle progression in 2F7 AIDS-related Burkitt's lymphoma cells at 14 µM
Catalog No. 50-291-3622 Supplier Cayman Chemical Supplier No. 354945 MG
A PKCβ Inhibitor (IC50s = 21 and 5 nM, respectively); selective for PKCβ1 and PKCβ2 over PKCα, PKCε, and PKCγ (IC50s = 331, 2,807, and >1,000 nM, respectively), as well as JAK1, JAK2, and TYK2 (IC50s = 770, 3,850, and 2,310 nM, respectively); inhibits JAK3 (IC50 = 17 nM); induces apoptosis and inhibits cell cycle progression in 2F7 AIDS-related Burkitt's lymphoma cells at 14 µM