Learn More
Sino Biological Recombinant Human PKC iota Protein (Full Length, GST Tag), Active
Catalog No. 502568135
Encompass_Preferred
Recombinant full length human PKC iota was expressed by baculovirus in Sf9 insect cells using a N-terminal GST tag.
Catalog No. 50-256-8135 Supplier Sino Biological Supplier No. P6810G10
Recombinant full length human PKC iota was expressed by baculovirus in Sf9 insect cells using a N-terminal GST tag.