Learn More
Eurofins Discoverx PKC iota, active, 10 µg. (14-505)
Supplier:  Eurofins Discoverx 14505
Encompass
N-terminal 6xHis-tagged recombinant, human full length PKC iota PKC Lipid activator (Catalog # 20-133) is required for use. For detailed product information, visit www.discoverx.com/proteins.
Catalog No. 50-199-8883