Learn More
Medchemexpress LLC Protein kinase C iota, human, recombinant (Sf9), His-tagged | 100UG
SDP
Catalog No. 5000175532
Encompass_Preferred
Recombinant human protein kinase C iota (PRKCI) expressed in Sf9 insect cells and supplied as a filtered, buffered solution suitable for biochemical and structural studies. The preparation includes stabilizing agents and protease inhibitors to help preserve protein integrity during storage and handling.
  • Expressed in Sf9 insect cells.
  • His-tagged recombinant human protein (amino acids M10-V596).
  • Approximate molecular weight 90-110 kDa.
  • Supplied in 50 mM Tris, 100 mM NaCl, 0.5 mM GSH, 0.5 mM PMSF, 10% glycerol, pH 7.4.
  • 0.22 μm filtered solution.
  • Store at -80°C; stable at -20°C for short-term use; avoid repeated freeze-thaw cycles.
  • Suitable for biochemical assays and structural studies.
Catalog No. 50-001-75532 Supplier Medchemexpress LLC Supplier No. HYP702620100UG
Recombinant human protein kinase C iota (PRKCI) expressed in Sf9 insect cells and supplied as a filtered, buffered solution suitable for biochemical and structural studies. The preparation includes stabilizing agents and protease inhibitors to help preserve protein integrity during storage and handling.
  • Expressed in Sf9 insect cells.
  • His-tagged recombinant human protein (amino acids M10-V596).
  • Approximate molecular weight 90-110 kDa.
  • Supplied in 50 mM Tris, 100 mM NaCl, 0.5 mM GSH, 0.5 mM PMSF, 10% glycerol, pH 7.4.
  • 0.22 μm filtered solution.
  • Store at -80°C; stable at -20°C for short-term use; avoid repeated freeze-thaw cycles.
  • Suitable for biochemical assays and structural studies.