Learn More
Medchemexpress LLC PRKCI protein, human recombinant (Sf9, His tag) | 10UG
SDP
Catalog No. 5000175794
Encompass_Preferred
Recombinant human PRKCI protein expressed in Sf9 insect cells and purified with a His tag. Supplied as a 0.22 μm filtered solution in a Tris-based buffer with glycerol and stabilizers, the protein is intended for biochemical assays, signaling studies, and reagent validation. Ship on dry ice; store at -80°C for long-term stability and avoid repeated freeze-thaw cycles.
  • Recombinant human protein expressed in Sf9 insect cells
  • His tag for straightforward purification and detection
  • Low endotoxin level (<1 EU/μg) suitable for sensitive assays
  • Provided in Tris buffer with NaCl, GSH, PMSF, and 10% glycerol for stability
  • Shipped on dry ice and stable at -80°C for long-term storage
Catalog No. 50-001-75794 Supplier Medchemexpress LLC Supplier No. HYP70262010UG
Recombinant human PRKCI protein expressed in Sf9 insect cells and purified with a His tag. Supplied as a 0.22 μm filtered solution in a Tris-based buffer with glycerol and stabilizers, the protein is intended for biochemical assays, signaling studies, and reagent validation. Ship on dry ice; store at -80°C for long-term stability and avoid repeated freeze-thaw cycles.
  • Recombinant human protein expressed in Sf9 insect cells
  • His tag for straightforward purification and detection
  • Low endotoxin level (<1 EU/μg) suitable for sensitive assays
  • Provided in Tris buffer with NaCl, GSH, PMSF, and 10% glycerol for stability
  • Shipped on dry ice and stable at -80°C for long-term storage