Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Protein kinase C iota | 50UG
SDP

Catalog No. 5000176635
Encompass_Preferred

PRKCI Protein, Human (sf9, His) is a recombinant human protein kinase C iota produced in Sf9 insect cells and supplied with a His tag for research use in biochemical and cell-based assays. The protein is delivered as a filtered solution, has an approximate molecular weight of 90-110 kDa, and should be stored at -80°C with minimal freeze-thaw cycles.

  • Recombinant human protein produced in Sf9 insect cells.
  • C-terminal His tag for purification and detection.
  • Supplied as a 0.22 μm filtered solution to reduce contamination.
  • Molecular weight approximately 90-110 kDa.
  • Endotoxin level <1 EU/μg.
  • Store at -80°C and avoid repeated freeze-thaw cycles.

Catalog No. 50-001-76635 Supplier Medchemexpress LLC Supplier No. HYP70262050UG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

PRKCI Protein, Human (sf9, His) is a recombinant human protein kinase C iota produced in Sf9 insect cells and supplied with a His tag for research use in biochemical and cell-based assays. The protein is delivered as a filtered solution, has an approximate molecular weight of 90-110 kDa, and should be stored at -80°C with minimal freeze-thaw cycles.

  • Recombinant human protein produced in Sf9 insect cells.
  • C-terminal His tag for purification and detection.
  • Supplied as a 0.22 μm filtered solution to reduce contamination.
  • Molecular weight approximately 90-110 kDa.
  • Endotoxin level <1 EU/μg.
  • Store at -80°C and avoid repeated freeze-thaw cycles.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.