Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC PRKCI protein, human (sf9, His) | 5UG
SDP

Catalog No. 5000175177
Encompass_Preferred

Recombinant human PRKCI (protein kinase C iota) produced in Sf9 insect cells and purified with a His tag. Supplied as a 0.22 μm filtered solution in a Tris-based buffer with glycerol; reported molecular weight is 90-110 kDa and endotoxin level is <1 EU/μg. Store frozen for long-term stability and avoid repeated freeze-thaw cycles.

  • Recombinant human protein expressed in Sf9 insect cells.
  • His-tagged for affinity purification and detection.
  • Molecular weight 90-110 kDa by SDS-PAGE.
  • Endotoxin level <1 EU/μg (LAL method).
  • Supplied in 50 mM Tris, 100 mM NaCl, 0.5 mM GSH, 0.5 mM PMSF, 10% glycerol, pH 7.4.
  • Store at -80°C for up to 1 year; avoid repeated freeze-thaw.

Catalog No. 50-001-75177 Supplier Medchemexpress LLC Supplier No. HYP7026205UG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Recombinant human PRKCI (protein kinase C iota) produced in Sf9 insect cells and purified with a His tag. Supplied as a 0.22 μm filtered solution in a Tris-based buffer with glycerol; reported molecular weight is 90-110 kDa and endotoxin level is <1 EU/μg. Store frozen for long-term stability and avoid repeated freeze-thaw cycles.

  • Recombinant human protein expressed in Sf9 insect cells.
  • His-tagged for affinity purification and detection.
  • Molecular weight 90-110 kDa by SDS-PAGE.
  • Endotoxin level <1 EU/μg (LAL method).
  • Supplied in 50 mM Tris, 100 mM NaCl, 0.5 mM GSH, 0.5 mM PMSF, 10% glycerol, pH 7.4.
  • Store at -80°C for up to 1 year; avoid repeated freeze-thaw.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.