Learn More
TARGETMOL CHEMICALS INC PKC-iota inhibitor 1 1MG
SDP
Catalog No. 502878085
Encompass_Preferred
Also available in 1 mL, 2 mg, 5 mg, 10 mg, 25 mg, 50 mg, 100 mg, 500 mg and bulk. Please contact Fisher for quotes. PKC-iota inhibitor 1 is an inhibitor of protein kinase C-iota (PKC-ι ; IC50 0.34 uM) Purity 98.82%
Catalog No. 50-287-8085 Supplier TARGETMOL CHEMICALS INC Supplier No. T87641MG
Also available in 1 mL, 2 mg, 5 mg, 10 mg, 25 mg, 50 mg, 100 mg, 500 mg and bulk. Please contact Fisher for quotes. PKC-iota inhibitor 1 is an inhibitor of protein kinase C-iota (PKC-ι ; IC50 0.34 uM) Purity 98.82%