Learn More
Medchemexpress LLC PKC-iota inhibitor 1 | 2328094-11-5 | 99.6% | 374.44 g·mol⁻¹ | C21H22N6O | 1MG
SDP
Catalog No. 5000239643
Encompass_Preferred
PKC-iota inhibitor 1 is a small-molecule inhibitor of protein kinase C-iota (PKC-ι) characterized by an IC50 of 0.34 μM. It is provided as a high-purity research chemical intended for biochemical and cellular studies, including oncology-focused assays. Product documentation includes a data sheet and a safety data sheet for laboratory handling and risk information.
  • Small-molecule PKC-ι inhibitor with an IC50 of 0.34 μM.
  • High purity (99.6%).
  • Molecular formula C21H22N6O and molecular weight 374.44 g·mol⁻¹.
  • Available in multiple vial sizes for research (1 mg, 5 mg, 10 mg, 50 mg, 100 mg).
  • Comes with a product data sheet and safety data sheet for handling guidance.
Catalog No. 50-002-39643 Supplier Medchemexpress LLC Supplier No. HY1261461MG
PKC-iota inhibitor 1 is a small-molecule inhibitor of protein kinase C-iota (PKC-ι) characterized by an IC50 of 0.34 μM. It is provided as a high-purity research chemical intended for biochemical and cellular studies, including oncology-focused assays. Product documentation includes a data sheet and a safety data sheet for laboratory handling and risk information.
  • Small-molecule PKC-ι inhibitor with an IC50 of 0.34 μM.
  • High purity (99.6%).
  • Molecular formula C21H22N6O and molecular weight 374.44 g·mol⁻¹.
  • Available in multiple vial sizes for research (1 mg, 5 mg, 10 mg, 50 mg, 100 mg).
  • Comes with a product data sheet and safety data sheet for handling guidance.