Learn More
Medchemexpress LLC PKC-iota inhibitor 1 | 2328094-11-5 | MFCD32062834 | 99.6% | 374.44 g/mol | C21H22N6O | 10MM 1ML
SDP
Catalog No. 5000239275
Encompass_Preferred
PKC-iota inhibitor 1 is a small-molecule research reagent that inhibits protein kinase C iota (PKC-ι) with an IC50 of 0.34 μM. It is supplied as a ready-to-use 10 mM solution in DMSO or as solid powder in multiple sizes, and is intended for biochemical and cellular research; safety data sheet and product datasheet are provided.
Catalog No. 50-002-39275 Supplier Medchemexpress LLC Supplier No. HY12614610MM 1ML
PKC-iota inhibitor 1 is a small-molecule research reagent that inhibits protein kinase C iota (PKC-ι) with an IC50 of 0.34 μM. It is supplied as a ready-to-use 10 mM solution in DMSO or as solid powder in multiple sizes, and is intended for biochemical and cellular research; safety data sheet and product datasheet are provided.