Learn More
Sino Biological Assay Buffer and Co-factors: PKC Lipid Activator, 10x
Catalog No. 502568740
Encompass_Preferred
10x stock solution used for assaying protein kinase activity of PKC family members. Vortex or sonicate prior to use.
Catalog No. 50-256-8740 Supplier Sino Biological Supplier No. L5139500
10x stock solution used for assaying protein kinase activity of PKC family members. Vortex or sonicate prior to use.