Learn More
Enzo Life Sciences PKC mu (human), (recombinant) (GST-tag) (5 µg)
SDP
Supplier:  Enzo Life Sciences BMLSE3480005
Encompass_Preferred
Alternative name: Protein kinase C mu, Protein kinase D, PKD. Source: Produced in Sf9 insect cells. Active, full length protein kinase C mu (PKCµ, protein kinase D, PKD) is fused at the N-terminus to a GST-tag. Produced in a baculovirus expression system. Long Term Storage: -80°C.
Catalog No. 50-201-1992