Learn More
Enzo Life Sciences PKC polyclonal antibody (100µg)
SDP
Supplier:  Enzo Life Sciences ADIKAPPK003E
Encompass_Preferred
Host: Rabbit. Immunogen: Synthetic peptide corresponding to a portion of human PKC. The sequence is completely conserved between the α, β, and δ isoforms of PKC. UniProt ID: P17252 (PKCalpha). Source: Purified from rabbit serum. Species reactivity: Human, Mouse, Rat, Sheep, Rabbit, Porcine, Monkey, Guinea pig, Dog, Chicken, Bovine. Applications: IHC, IP, WB. Recommended Dilutions: Immunohistochemistry (1:50) Immunoprecipitation (12.5µg/ml) Western Blot (2µg/ml, ECL) Suggested dilutions/conditions may not be available for all applications. Optimal conditions must be determined individually for each application. Purity: Protein A affinity purified. Formulation: Liquid. In borate buffered saline, pH 8.2.
Catalog No. 50-200-7288