Learn More
Medchemexpress LLC PKC/PKD-IN-1 (Compound 13C) | 1071135-06-2 | 98.4% | 388.51 g·mol⁻¹ | C23H28N6 | 5 MG
SDP
Catalog No. 5000264485
Encompass_Preferred
PKC/PKD-IN-1 is an orally active dual protein kinase C/D inhibitor used in preclinical research; it inhibits PKD1 with an IC50 of 0.6 nM and has been used to study cardiac hypertrophy and heart failure. For research use only.
  • Potent PKD1 inhibition (IC50 0.6 nM).
  • Orally active compound suitable for in vivo studies.
  • High purity supporting reproducible results.
  • Available in small mg-scale quantities for dosing studies.
  • Supplied with supporting datasheet for assay planning.
Catalog No. 50-002-64485 Supplier Medchemexpress LLC Supplier No. HY1281425MG
PKC/PKD-IN-1 is an orally active dual protein kinase C/D inhibitor used in preclinical research; it inhibits PKD1 with an IC50 of 0.6 nM and has been used to study cardiac hypertrophy and heart failure. For research use only.
  • Potent PKD1 inhibition (IC50 0.6 nM).
  • Orally active compound suitable for in vivo studies.
  • High purity supporting reproducible results.
  • Available in small mg-scale quantities for dosing studies.
  • Supplied with supporting datasheet for assay planning.