Learn More
Cayman Chemical PKC Pseudosubstrate InhIbIto
SDP
Supplier:  Cayman Chemical 17482500 UG
Encompass_Preferred
A synthetic peptide that that selectively, reversibly, and substrate-competitively inhibits PKCζ activity and, thus, is used to delineate the signaling functions of PKCζ
Catalog No. 50-296-7863