Learn More
Cayman Chemical PKCζ Pseudosubstrate Inhibitor, Purity: ≥95%, CAS: 799764-07-1, 5 mg
SDP
Catalog No. 502965049
Encompass_Preferred
A synthetic peptide that that selectively, reversibly, and substrate-competitively inhibits PKCζ activity and, thus, is used to delineate the signaling functions of PKCζ
Catalog No. 50-296-5049 Supplier Cayman Chemical Supplier No. 174825 MG
A synthetic peptide that that selectively, reversibly, and substrate-competitively inhibits PKCζ activity and, thus, is used to delineate the signaling functions of PKCζ