Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PKC Sampler Kit, BD

Catalog No. BDB611421 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
10 μg (Each)
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB611421 10 μg (Each)
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB611421 Supplier BD Biosciences Supplier No. 611421
Add to Cart
Add to Cart

Host Species:
Isotype: IgG1, IgG2a, IgG2b, IgG1, IgM
Cross Reactivity Species: Hu, Rt

Western Blotting

TRUSTED_SUSTAINABILITY

Specifications

Conjugate Unconjugated
Purification Method Affinity Purified
Quantity 10 μg (Each)
Isotype IgG1, IgG2a, IgG2b, IgG1, IgM
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.