Learn More
Cayman Chemical PKCδ Substrate (trifluoroacetate salt), Purity: ≥98%, 10 mg
SDP
Catalog No. 502916188
Encompass_Preferred
A peptide substrate for PKCδ; has been used to monitor PKCδ activity in cell-free and cell-based assays
Catalog No. 50-291-6188 Supplier Cayman Chemical Supplier No. 3751310 MG
A peptide substrate for PKCδ; has been used to monitor PKCδ activity in cell-free and cell-based assays