Learn More
Medchemexpress LLC PKC-theta inhibitor 1 | 1160501-81-4 | 99.7% | 348.32 g·mol⁻¹ | C17H15F3N4O | 5MG
SDP
Catalog No. 5000236931
Encompass_Preferred
PKC-theta inhibitor 1 is an orally active, selective ATP-competitive inhibitor of protein kinase C theta (PKCθ) used in biochemical and cellular research to probe PKCθ-mediated signaling pathways. It is a small-molecule research reagent with reported high potency and listed high purity.
  • Selective ATP-competitive inhibition of PKCθ for target-specific studies.
  • High potency with reported Ki ≈ 6 nM.
  • High purity (99.7%) suitable for biochemical assays.
  • Characterized small molecule with formula C17H15F3N4O and molecular weight 348.32 g·mol⁻¹.
  • Available in multiple research pack sizes for assay flexibility.
Catalog No. 50-002-36931 Supplier Medchemexpress LLC Supplier No. HY1263285MG
PKC-theta inhibitor 1 is an orally active, selective ATP-competitive inhibitor of protein kinase C theta (PKCθ) used in biochemical and cellular research to probe PKCθ-mediated signaling pathways. It is a small-molecule research reagent with reported high potency and listed high purity.
  • Selective ATP-competitive inhibition of PKCθ for target-specific studies.
  • High potency with reported Ki ≈ 6 nM.
  • High purity (99.7%) suitable for biochemical assays.
  • Characterized small molecule with formula C17H15F3N4O and molecular weight 348.32 g·mol⁻¹.
  • Available in multiple research pack sizes for assay flexibility.