Learn More
Medchemexpress LLC PKC-theta inhibitor 2 | 736048-65-0 | 99.9% | 454.45 | 10 MM 1 ML
SDP
Catalog No. 5000436993
Encompass_Preferred
PKC-theta inhibitor 2 is a potent and selective PKC-θ inhibitor with an IC50 value of 18 nM. It is a 2,4-diamino-5-nitropyrimidine compound with the 2-position substituted by o-trifluoromethoxy.
  • Potent and selective PKC-θ inhibitor.
  • For research use only.
Catalog No. 50-004-36993 Supplier Medchemexpress LLC Supplier No. HY11268110MM/1ML
Add to Cart
Add to Cart
PKC-theta inhibitor 2 is a potent and selective PKC-θ inhibitor with an IC50 value of 18 nM. It is a 2,4-diamino-5-nitropyrimidine compound with the 2-position substituted by o-trifluoromethoxy.
  • Potent and selective PKC-θ inhibitor.
  • For research use only.