Learn More
Medchemexpress LLC PKC-theta inhibitor 2 | 736048-65-0 | 99.9% | 454.45 | 100 MG
SDP
Catalog No. 5000438478
Encompass_Preferred
PKC-theta inhibitor 2 is a potent and selective inhibitor of PKC-θ, exhibiting an IC50 value of 18 nM. This compound is identified as a 2,4-diamino-5-nitropyrimidine, characterized by a trifluoromethoxy substitution at the 2-position.
  • Potent and selective inhibition of PKC-θ.
  • Exhibits an IC50 value of 18 nM.
  • Characterized as a 2,4-diamino-5-nitropyrimidine compound.
  • Features an o-trifluoromethoxy substitution at the 2-position.
Catalog No. 50-004-38478 Supplier Medchemexpress LLC Supplier No. HY112681100MG
PKC-theta inhibitor 2 is a potent and selective inhibitor of PKC-θ, exhibiting an IC50 value of 18 nM. This compound is identified as a 2,4-diamino-5-nitropyrimidine, characterized by a trifluoromethoxy substitution at the 2-position.
  • Potent and selective inhibition of PKC-θ.
  • Exhibits an IC50 value of 18 nM.
  • Characterized as a 2,4-diamino-5-nitropyrimidine compound.
  • Features an o-trifluoromethoxy substitution at the 2-position.