Learn More
Medchemexpress LLC PKC-theta inhibitor 2 | 736048-65-0 | 99.9% | 454.45 | 50 MG
SDP
Catalog No. 5000435672
Encompass_Preferred
PKC-theta inhibitor 2 is a potent and selective PKC-θ inhibitor with an IC50 value of 18 nM. It is a 2,4-diamino-5-nitropyrimidine compound with 2-position substituted by o-trifluoromethoxy.
  • Potent and selective PKC-θ inhibitor
  • Targets PKC-θ
  • Inhibits epigenetics and TGF-beta/Smad pathways
  • Soluble in DMSO at 62.5 mg/mL (137.53 mM)
  • Multiple in vivo solubility options available
Catalog No. 50-004-35672 Supplier Medchemexpress LLC Supplier No. HY11268150MG
PKC-theta inhibitor 2 is a potent and selective PKC-θ inhibitor with an IC50 value of 18 nM. It is a 2,4-diamino-5-nitropyrimidine compound with 2-position substituted by o-trifluoromethoxy.
  • Potent and selective PKC-θ inhibitor
  • Targets PKC-θ
  • Inhibits epigenetics and TGF-beta/Smad pathways
  • Soluble in DMSO at 62.5 mg/mL (137.53 mM)
  • Multiple in vivo solubility options available