Learn More
Selleck Chemical LLC PKC-theta inhibitor
SDP
Catalog No. 5000002788
Encompass_Preferred
PKC-theta inhibitor (compound 20) inhibits PKC- with an IC50 of 18 nM
Catalog No. 50-000-02788 Supplier Selleck Chemical LLC Supplier No. S6577100MG
PKC-theta inhibitor (compound 20) inhibits PKC- with an IC50 of 18 nM