Learn More
Enzo Life Sciences PKC zeta (113-125) (myristoylated) (0.5mg)
SDP
Supplier:  Enzo Life Sciences BMLP2190500
Encompass_Preferred
Pseudosubstrate sequence from human PKCζ (113-125) completely conserved in rabbit, rat, mouse and Xenopus. Sequence is also identical in human PKCι; (114-126) and mouse PKCλ. The N-terminus is myristoylated to allow cell permeability. Peptide inhibitor of atypical PKC’s (calcium/phorbol ester/diacyglycerol insensitive). Useful in conjunction with the Myristoylated PKC (20-28), Cell-permeable PKC Inhibitor (BML-P205). Alternative name: Protein kinase C zeta (113-125) (myristoylated), PKC zeta pseudosubstrate. Purity: ≥97% (HPLC). Formulation: Lyophilized. Solubility: Soluble in water or DMSO. Long Term Storage: -20°C.
Catalog No. 50-201-1293