Learn More
Enzo Life Sciences PKC zeta (human), (recombinant) (GST-tag) (5 µg)
SDP
Supplier:  Enzo Life Sciences BMLSE4130005
Encompass_Preferred
Alternative name: Protein kinase C zeta. Formulation: Liquid. In 50mM TRIS-HCl, pH 7.5, containing 150mM NaCl, 10mM glutathione, 0.1mM EDTA, 0.25mM DTT, 0.1mM PMSF and 25% glycerol. Source: Produced in insect cells. Produced in a baculovirus expression system. Long Term Storage: -80°C.
Catalog No. 50-201-2016