Learn More
Enzo Life Sciences PKC zeta pseudosubstrate (myristoylated) (1mg)
SDP
Catalog No. 502008360
Encompass_Preferred
Selective cell permeable inhibitor of PKCζ. Activates eNOS (NOS III) through Akt phosphorylation and can be considered as a nitric oxide production activator in endothelial cells. Alternative name: Protein kinase C zeta pseudosubstrate, PKC zeta (113-125) (myristoylated). Formula: C90H154N30O17. MW: 1928.4. Purity: ≥95% (HPLC). Appearance: White to off-white powder. Formulation: Counter Ion: TFA. Peptide content: ≥60%. Solubility: Soluble in water, DMSO or water-methanol (50:50) . Long Term Storage: -20°C.
Catalog No. 50-200-8360 Supplier Enzo Life Sciences Supplier No. ALX260155M001
Selective cell permeable inhibitor of PKCζ. Activates eNOS (NOS III) through Akt phosphorylation and can be considered as a nitric oxide production activator in endothelial cells. Alternative name: Protein kinase C zeta pseudosubstrate, PKC zeta (113-125) (myristoylated). Formula: C90H154N30O17. MW: 1928.4. Purity: ≥95% (HPLC). Appearance: White to off-white powder. Formulation: Counter Ion: TFA. Peptide content: ≥60%. Solubility: Soluble in water, DMSO or water-methanol (50:50) . Long Term Storage: -20°C.