Learn More
Enzo Life Sciences PKC zeta substrate (1mg)
SDP
Catalog No. 502011264
Encompass_Preferred
This peptide sequence, RRFKRQGSFFYFF, was selected from a peptide library for optimal phosphorylation by protein kinase C ζ (PKCζ). Alternative name: Protein kinase C zeta substrate. Long Term Storage: -20°C.
Catalog No. 50-201-1264 Supplier Enzo Life Sciences Supplier No. BMLP1560001
This peptide sequence, RRFKRQGSFFYFF, was selected from a peptide library for optimal phosphorylation by protein kinase C ζ (PKCζ). Alternative name: Protein kinase C zeta substrate. Long Term Storage: -20°C.