Learn More
Cayman Chemical PKC0 InhIbItrhydrochLRIde 10mg
SDP
Catalog No. 502912862
Encompass_Preferred
A PKCθ inhibitor (IC50 = 18 nM)
Catalog No. 50-291-2862 Supplier Cayman Chemical Supplier No. 3616510 MG
A PKCθ inhibitor (IC50 = 18 nM)