Learn More
Sigma Aldrich Fine Chemicals Biosciences PKCalpha Protein active 10
Catalog No. 502905873
Encompass
Recombinant full-length human PKCalpha containing N-terminal His6 tagPKC Lipid activator (Catalog 20-133) is required for use.
Catalog No. 50-290-5873 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. 14484
Recombinant full-length human PKCalpha containing N-terminal His6 tagPKC Lipid activator (Catalog 20-133) is required for use.