Learn More
Carnabio Usa Inc PKCB1(PRKCB1)/10UG/FULL-LENGTH
Supplier:  Carnabio Usa Inc 0113410
Encompass
PKC1(PRKCB1), Full-length, Wild type, Amino Acid 1-671, NP_997700.1, Expressed in Insect (sf21); 10microg
Catalog No. 50-754-0942