Learn More
Eurofins Discoverx PKCβI, active, 250 µg. (14-503M)
Supplier:  Eurofins Discoverx 14503M
Encompass
N-terminal 6xHis-tagged recombinant, human PKCβ amino acids 2-end. PKC Lipid activator (Catalog # 20-133) is required for use. For detailed product information, visit www.discoverx.com/proteins.
Catalog No. 50-199-8880