Learn More
Carnabio Usa Inc PKCD(PRKCD)/5UG/FULL-LENGTH
Catalog No. 507540585
Encompass
PKC(PRKCD), Full-length, Wild type, Amino Acid 1-676, NP_006245.2, Expressed in Insect (sf21); 5microg
Catalog No. 50-754-0585 Supplier Carnabio Usa Inc Supplier No. 011355
PKC(PRKCD), Full-length, Wild type, Amino Acid 1-676, NP_006245.2, Expressed in Insect (sf21); 5microg