Learn More
Carnabio Usa Inc PKCG(PRKCG)/10UG/FULL-LENGTH
Catalog No. 507540944
Encompass
PKC(PRKCG), Full-length, Wild type, Amino Acid 1-697, NP_002730.1, Expressed in Insect (sf21); 10microg
Catalog No. 50-754-0944 Supplier Carnabio Usa Inc Supplier No. 0113710
PKC(PRKCG), Full-length, Wild type, Amino Acid 1-697, NP_002730.1, Expressed in Insect (sf21); 10microg