Learn More
Carnabio Usa Inc PKCI(PRKCI)/5UG/FULL-LENGTH
Supplier:  Carnabio Usa Inc 011395
Encompass
PKC(PRKCI), Full-length, Wild type, Amino Acid 1-587, NM_002740, Expressed in Insect (sf21); 5microg
Catalog No. 50-754-0590