Learn More
TARGETMOL CHEMICALS INC PKCIOTA-IN-2 5MG
SDP
Catalog No. 502578948
Encompass_Preferred
Also available in 1 mg 2 mg 10 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. PKCiota-IN-2 (PKCiota-IN-49) is a selective and potent PKCiota (PKC-(iota)) inhibitor with an IC50 of 2.8 nM. PKCiota-IN-2 also inhibits PKC-(alpha) and PKC-(epsilon) with IC50s of 71 nM and 350 nM respectivelypurity: 99%
Catalog No. 50-257-8948 Supplier TARGETMOL CHEMICALS INC Supplier No. T98635MG
Also available in 1 mg 2 mg 10 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. PKCiota-IN-2 (PKCiota-IN-49) is a selective and potent PKCiota (PKC-(iota)) inhibitor with an IC50 of 2.8 nM. PKCiota-IN-2 also inhibits PKC-(alpha) and PKC-(epsilon) with IC50s of 71 nM and 350 nM respectivelypurity: 99%