Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC PKCiota-IN-2 formic | 2230055-52-2 | 99.0% | 402.36 | 25 MG
SDP

Catalog No. 5000437620
Encompass_Preferred

PKCiota-IN-2 formic is a potent PKCiota (PKC-ι) inhibitor with an IC50 of 2.8 nM. It also inhibits PKC-α and PKC-ε with IC50s of 71 nM and 350 nM, respectively. This product is for research use only.

  • Appearance: Solid
  • Color: Light yellow to yellow
  • Shipping: Room temperature in continental US; may vary elsewhere
  • Storage: 4°C, sealed storage, away from moisture and light. In solvent, store at -80°C for 6 months or -20°C for 1 month (sealed, away from moisture and light).
  • Solubility (in vitro): DMSO: 100 mg/mL (248.53 mM; requires ultrasonic).

Catalog No. 50-004-37620 Supplier Medchemexpress LLC Supplier No. 25MGHY122858A
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

PKCiota-IN-2 formic is a potent PKCiota (PKC-ι) inhibitor with an IC50 of 2.8 nM. It also inhibits PKC-α and PKC-ε with IC50s of 71 nM and 350 nM, respectively. This product is for research use only.

  • Appearance: Solid
  • Color: Light yellow to yellow
  • Shipping: Room temperature in continental US; may vary elsewhere
  • Storage: 4°C, sealed storage, away from moisture and light. In solvent, store at -80°C for 6 months or -20°C for 1 month (sealed, away from moisture and light).
  • Solubility (in vitro): DMSO: 100 mg/mL (248.53 mM; requires ultrasonic).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.