Learn More
Medchemexpress LLC Pkciota-IN-2 formic | 99.0% | 402.36 | C25H23N5O3 | 5 MG
SDP
Catalog No. 5000264538
Encompass_Preferred
PKCiota-IN-2 formic is a research-grade small-molecule inhibitor targeting atypical protein kinase C iota (PKCι). It exhibits low-nanomolar potency and a defined selectivity profile, and is supplied as a powder for laboratory use with recommended sealed, refrigerated storage.
  • Low-nanomolar potency against PKCι (IC50 2.8 nM).
  • Inhibits PKCα and PKCε with higher IC50s (71 nM and 350 nM).
  • High chemical purity (99.04%).
  • Molecular weight 402.36 g·mol⁻¹.
  • Provided as a powder with refrigerated storage recommended.
  • Available in small research pack sizes (including 5 mg).
Catalog No. 50-002-64538 Supplier Medchemexpress LLC Supplier No. HY122858A5MG
PKCiota-IN-2 formic is a research-grade small-molecule inhibitor targeting atypical protein kinase C iota (PKCι). It exhibits low-nanomolar potency and a defined selectivity profile, and is supplied as a powder for laboratory use with recommended sealed, refrigerated storage.
  • Low-nanomolar potency against PKCι (IC50 2.8 nM).
  • Inhibits PKCα and PKCε with higher IC50s (71 nM and 350 nM).
  • High chemical purity (99.04%).
  • Molecular weight 402.36 g·mol⁻¹.
  • Provided as a powder with refrigerated storage recommended.
  • Available in small research pack sizes (including 5 mg).