Learn More
Medchemexpress LLC PKCiota-IN-2 formic | 99.04% | 402.36 | 50 MG
SDP
Catalog No. 5000437811
Encompass_Preferred
PKCiota-IN-2 formic is a potent inhibitor of PKCiota (PKC-ι), with an IC50 of 2.8 nM. It also demonstrates inhibitory activity against PKC-α and PKC-ε, exhibiting IC50s of 71 nM and 350 nM, respectively. This product is intended for research use only and is not for sale to patients.
  • Potent inhibition of PKCiota.
  • Exhibits inhibitory effects on PKC-alpha and PKC-epsilon.
  • Available in various quantities, including solid and solution forms.
Catalog No. 50-004-37811 Supplier Medchemexpress LLC Supplier No. HY122858A50MG
PKCiota-IN-2 formic is a potent inhibitor of PKCiota (PKC-ι), with an IC50 of 2.8 nM. It also demonstrates inhibitory activity against PKC-α and PKC-ε, exhibiting IC50s of 71 nM and 350 nM, respectively. This product is intended for research use only and is not for sale to patients.
  • Potent inhibition of PKCiota.
  • Exhibits inhibitory effects on PKC-alpha and PKC-epsilon.
  • Available in various quantities, including solid and solution forms.