Learn More
Sino Biological Peptide Substrates: PKCtide
Catalog No. 502568357
Encompass_Preferred
The PKCtide peptide sequence (ERMRPRKRQGSVRRRV) is based on protein kinase C epsilon (amino acid 149-164).
Catalog No. 50-256-8357 Supplier Sino Biological Supplier No. P1558
The PKCtide peptide sequence (ERMRPRKRQGSVRRRV) is based on protein kinase C epsilon (amino acid 149-164).