Learn More
Eurofins Discoverx PKCγ, active, 250 µg. (14-483M)
Supplier:  Eurofins Discoverx 14483M
Encompass
N-terminal 6xHis-tagged recombinant, human full length PKCγ. PKC Lipid activator (Catalog # 20-133) is required for use. For detailed product information, visit www.discoverx.com/proteins.
Catalog No. 50-199-8856