Learn More
Carnabio Usa Inc PKD3(PRKD3)/5UG/FULL-LENGTH
Supplier:  Carnabio Usa Inc 021595
Encompass
PKD3(PRKD3), Full-length, Wild type, Amino Acid 1-890, NP_005804.1, Expressed in Insect (sf21); 5microg
Catalog No. 50-754-0593