Learn More
Beckman Coulter 96 Well-Plate for use with Vi-CELL BLU (50 count)
Catalog No. NC2740176 Shop All Beckman Coulter Products
Encompass_Preferred
Quantity of 50, 96-well plates for use with Vi-CELL BLU cell viability analyzer. Use the plate cover slip, C24842, with the plate.Need a quantity of 5? Click here for the C99716
Catalog No. NC2740176 Supplier Beckman Coulter Supplier No. C99715
Quantity of 50, 96-well plates for use with Vi-CELL BLU cell viability analyzer. Use the plate cover slip, C24842, with the plate.Need a quantity of 5? Click here for the C99716