Learn More
Medchemexpress LLC HY-112197 5mg Medchemexpress, PKG drug G1 CAS:374703-78-3 Purity:>98%
SDP
Catalog No. 501969058
Encompass_Preferred
Medchemexpress, HY-112197 5mg PKG drug G1 CAS:374703-78-3 PKG drug G1 targets C42 of PKG Iα. PKG drug G1 can couple to vasodilation and blood pressure lowering by a C42 PKG Iα-independent mechanism. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-9058 Supplier Medchemexpress LLC Supplier No. HY1121975MG
Medchemexpress, HY-112197 5mg PKG drug G1 CAS:374703-78-3 PKG drug G1 targets C42 of PKG Iα. PKG drug G1 can couple to vasodilation and blood pressure lowering by a C42 PKG Iα-independent mechanism. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.