Learn More
Medchemexpress LLC PKG inhibitor peptide | 82801-73-8 | 99.8% | 943.11 | 10 MG
SDP
Catalog No. 5000424491
Encompass_Preferred
PKG inhibitor peptide is an ATP-competitive inhibitor of cGMP-dependent protein kinase (PKG), with a Ki of 86 μM. This peptide is for research use only and not intended for patient use. It is provided as a white to off-white solid with a purity of 99.8%.
  • ATP-competitive inhibitor of cGMP-dependent protein kinase (PKG)
  • Ki of 86 μM (PKG)
  • Purity of 99.8%
  • White to off-white solid appearance
  • Stable under recommended storage conditions
  • For research use only
Catalog No. 50-004-24491 Supplier Medchemexpress LLC Supplier No. HYP129210MG
PKG inhibitor peptide is an ATP-competitive inhibitor of cGMP-dependent protein kinase (PKG), with a Ki of 86 μM. This peptide is for research use only and not intended for patient use. It is provided as a white to off-white solid with a purity of 99.8%.
  • ATP-competitive inhibitor of cGMP-dependent protein kinase (PKG)
  • Ki of 86 μM (PKG)
  • Purity of 99.8%
  • White to off-white solid appearance
  • Stable under recommended storage conditions
  • For research use only