Learn More
Cayman Chemical PKG InhIbItrtrIfluoroACTA 10mg
SDP
Catalog No. 502968889
Encompass_Preferred
A specific cGMP-dependent PKG inhibitor (Ki = 86 µM); a nonphosphorylatable synthetic peptide analogous to a substrate corresponding to the serine-32 phosphorylation site in histone H2B
Catalog No. 50-296-8889 Supplier Cayman Chemical Supplier No. 1599510 MG
A specific cGMP-dependent PKG inhibitor (Ki = 86 µM); a nonphosphorylatable synthetic peptide analogous to a substrate corresponding to the serine-32 phosphorylation site in histone H2B