Learn More
Hach Company LCP Salt Bridge, Qty: 1
Supplier:  Hach Company 609765000001
Encompass
Field replaceable salt bridge has a ceramic inner junction, PVDF outer junction, Viton O-Ring, and contains binary fill solution. Only for use with LCP 1-1/2 Inch differential pH sensors.
Catalog No. 50-199-7461