Learn More
Enzo Life Sciences PKG polyclonal antibody (50µg)
SDP
Supplier:  Enzo Life Sciences ADIKAPPK005D
Encompass_Preferred
Host: Rabbit. Immunogen: Synthetic peptide corresponding to a portion of human PKG. UniProt ID: Q13976. Source: Purified from rabbit serum. Species reactivity: Human, Mouse, Rat Porcine. Applications: IHC (PS), WB. Recommended Dilutions: Western Blot (1:1,000, colorimetric) Suggested dilutions/conditions may not be available for all applications. Optimal conditions must be determined individually for each application. Purity: Protein A affinity purified. Formulation: Liquid. In PBS, pH 7.2, containing 50% glycerol and 0.09% sodium azide.
Catalog No. 50-200-7291