Learn More
Apexbio Technology LLC PKH26 Red Fluorescent Cell Linker Kit(Synonyms: PKH26 Cell Membrane Dye, PKH26 Fluorescent Cell Labeling Kit), 250T.
SDP
Catalog No. 5000061399
Encompass_Preferred
PKH26 Red Fluorescent Cell Linker Kit comprises the red fluorescent probe PKH26 designed specifically to label cellular membranes PKH26 integrates stably into lipid regions of cell membranes emitting red fluorescence and enabling clear visualization of cells Due to low cytotoxicity low nonspecific background fluorescence and minimal dye transfer to neighboring unlabeled cells PKH26 has broad utility in tracking cells both in vitro and in vivo Its fluorescence is evenly partitioned between daughter cells during proliferation diminishing with each round of cell division facilitating proliferation assays PKH26 is frequently employed alone or in combination with PKH67 for cell tracking studies and proliferation analyses
Catalog No. 50-000-61399 Supplier Apexbio Technology LLC Supplier No. K2410250
PKH26 Red Fluorescent Cell Linker Kit comprises the red fluorescent probe PKH26 designed specifically to label cellular membranes PKH26 integrates stably into lipid regions of cell membranes emitting red fluorescence and enabling clear visualization of cells Due to low cytotoxicity low nonspecific background fluorescence and minimal dye transfer to neighboring unlabeled cells PKH26 has broad utility in tracking cells both in vitro and in vivo Its fluorescence is evenly partitioned between daughter cells during proliferation diminishing with each round of cell division facilitating proliferation assays PKH26 is frequently employed alone or in combination with PKH67 for cell tracking studies and proliferation analyses